
Cyclohexylamine

Synonym(s):
Aminocyclohexane, Cyclohexylamine
Linear Formula:
C6H11NH2
CAS Number:
Molecular Weight:
99.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing