
Chloroquine diphosphate solution

Synonym(s):
Chloroquine diphosphate solution
Empirical Formula (Hill Notation):
C18H26ClN3 · 2H3PO4
CAS Number:
Molecular Weight:
515.86
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing