Skip to Content
Merck
Boc-L-prolinal 97%

Boc-L-prolinal

Boc-L-prolinal 97%
Synonym(s):
(S)-N-Boc-pyrrolidine-2-carboxaldehyde, N-(tert-Butoxycarbonyl)-L-prolinal
Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing