Skip to Content
Merck
Bis(benzonitrile)palladium(II) chloride 95%

Bis(benzonitrile)palladium(II) chloride

Bis(benzonitrile)palladium(II) chloride 95%
Synonym(s):
Benzonitrile, palladium complex, Bis(benzonitrile)dichloropalladium(II), Bis(benzonitrile)palladium(II) chloride, Bis(phenylnitrile)dichloropalladium, Dibenzonitrilepalladium dichloride, Dichlorobis(benzonitrile)palladium, Dichlorobis(benzonitrile)palladium(II), Dichlorobis(phenyl cyanide)palladium, Palladium dichloride bis(benzonitrile), Palladium(II) chloride bis(benzonitrile) complex
Linear Formula:
(C6H5CN)2PdCl2
CAS Number:
Molecular Weight:
383.57
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing