
Bambuterol hydrochloride

Synonym(s):
Dimethylcarbmic acid 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-1,3-phenylene ester
Linear Formula:
C18H29N3O5 · HCl
CAS Number:
Molecular Weight:
403.90
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing