
α-Spinasterol

Synonym(s):
α-Spinasterin, (3β,5α,22E)-Stigmasta-7,22-dien-3-ol, (E)-5α-Stigmasta-7,22-dien-3β-ol, 5α-Stigmasta-7,22-dien-3β-ol, Bessisterol, Hitodesterol, Spinasterol
Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing