
Anti-Phosphothreonine antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-Phosphothreonine
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing