
Anisomycin

Synonym(s):
(2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate, 2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate, Flagecidin
Empirical Formula (Hill Notation):
C14H19NO4
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing