
9-Methylacridine

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing