
7-Aminoactinomycin D

Synonym(s):
7-AAD
Empirical Formula (Hill Notation):
C62H87N13O16
CAS Number:
Molecular Weight:
1270.43
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing