
3,4-Dicaffeoylquinic acid

Synonym(s):
4,5-Di-O-caffeoylquinic acid
Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing