
2-Sulfobenzoic acid ammonium salt

Synonym(s):
2-Sulfobenzoic acid ammonium salt
Linear Formula:
2-(HOOC)C6H4SO3NH4
CAS Number:
Molecular Weight:
219.22
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing