
2-Hydroxy-4-(octyloxy)benzophenone

Linear Formula:
CH3(CH2)7OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
326.43
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3