
2,4-Dimethylpyridine

Synonym(s):
2,4-Lutidine, 2,4-Dimethylpyridine
Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing