95116
Dimethachlor Metabolite CGA369873 sodium salt
PESTANAL®, analytical standard
Synonym(s):
2-[(2,6-Dimethylphenyl)amino]-2-oxoethanesulfonic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H12NNaO4S
CAS Number:
Molecular Weight:
265.26
MDL number:
UNSPSC Code:
41116107
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
Assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
storage temp.
2-8°C
General description
For R&D, and Analytical use only. Not for drug, household or other uses
Application
Metabolite for water testing.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service