Skip to Content
Merck
All Photos(1)

Key Documents

794-0715

HPLC Analysis of Paraquat and Diquat on SUPELCOSIL LC-18 after SPE using ENVI-8 DSK

application for SPE, application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: drinking water adjusted to pH to 10.
sample preparation: ENVI-8 DSK, 47 mm (57172)
SPE tube/cartridge: ENVI-8 DSK SPE Disk, 47 mm (57172)
condition: 10 mL acetonitrile; 2 × 10 mL reagent water; 10 mL conditioning solvent A (5 g cetyl trimethyl ammonium bromide and 5 mL conc. ammonium hydroxide in 500 mL DI water, dilute to 1 L); (2 × 10 mL reagent water; 10 mL conditioning solvent B (10 g hexanesulfonic acid, sodium salt and 10 mL conc. ammonium hydroxide in 250 mL DI water, dilute to 500 mL))
sample addition: 250 mL water 100 mL/min
elution: 0.5-1.0 mL acetonitrile (to cover/solvate disk); 2 × 4 mL eluting solution (13.5 mL orthophosphoric acid and 10.3 mL diethylamine in 500 mL DI water, dilute to 1 L)
column: SUPELCOSIL LC-18, 15 cm × 4.6 mm I.D., 5 μm particles (58230-U)
mobile phase: 3.5 mL triethylamine and 1.0 g 1-hexane-sulfonic acid, sodium salt to 800 mL DI H2O add orthophosphoric acid to pH 2.5, dilute to 1 L
flow rate: 1.0 mL/min
column temp.: 35 °C
detector: photodiode array, diquat - 308 nm, paraquat - 257 nm
injection: 100 μL

suitability

application for HPLC
application for SPE

application(s)

environmental

Application

Refer to US EPA Method 549.1 for full details

Sample Pre-treatment:
250 mL drinking water. Adjust water sample to pH to 10.5 ± 0.2 with 10% sodium hydroxide or 10% hydrochloric acid

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

analytical column

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service