Skip to Content
Merck
All Photos(2)

Key Documents

57620-U

Rheodyne® Injector

Rheodyne 7725

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
eCl@ss:
42079206
NACRES:
SB.51

description

manual with MBB

feature

number of ports 6
positions 2

manufacturer/tradename

Rheodyne 7725

parameter

7000 psi max. pressure

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC

recommended

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service