MAK564
Triglyceride Quantification Kit
sufficient for 100 colorimetric or fluorometric tests
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12352200
Recommended Products
Quality Level
application(s)
cosmetics
food and beverages
detection method
colorimetric
fluorometric
relevant disease(s)
obesity; cardiovascular diseases; neurological disorders
storage temp.
−20°C
Related Categories
General description
Triglycerides are the main constituents of vegetable oil, animal fat, LDL and VLDL, and play an important role as transporters of fatty acids as well as an energy source. Triglyceride is broken down into fatty acids and glycerol, after which both can serve as substrates for energy producing and metabolic pathways. High blood levels of triglycerides are implicated in atherosclerosis, heart disease and stroke as well as in pancreatitis. The Triglyceride Quantification Kit provides a sensitive, easy assay to measure triglyceride concentration in a variety of samples.
Features and Benefits
- Simple and high-throughput assay for the quantitative determination of triglycerides.
- Flexibility of either colorimetric or fluorometric detection all in one kit.
- Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file
Suitability
Suitable for the detection of triglyceride in various biological samples such as serum, plasma, saliva, other biological fluids, and tissue and cell culture samples.
Principle
In this assay, triglycerides are converted to free fatty acids and glycerol. The glycerol is then oxidized to generate a colorimetric (570 nm)/fluorometric (ex = 535 nm/em = 587 nm) product. The kit is sensitive to detect 2 pmole–10 nmole (2–10,000 mM range) of triglyceride in various samples. The kit also detects monoglycerides and diglycerides.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service