Key Documents
EP5920747002
Adapter for Eppendorf® S-4x Universal Large Rotor
capacity, 92 x 1.5/2.0 mL tube, pack of 2
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
capacity 92 x 1.5/2.0 mL tube
Holds 1.5/2.0 mL tubes
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5920747002 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with Universal-Large bucket
General description
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG