Skip to Content
Merck
All Photos(1)

Key Documents

RED.1

85092601, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RED.1, 85092601

biological source

mouse blood

growth mode

Suspension

karyotype

Aneuploid

morphology

Lymphoblast

products

Rauscher virus

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse leukaemic lymphoblast

Cell Line Description

A Rauscher virus induced erythroid leukaemia cell line. It is known to produce low amounts of Rauscher virus (a murine leukaemia virus).

Application

Tumourigenicity and virus studies: Rauscher virus

Culture Medium

DMEM or RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Let the cell culture media become acidic before sub-culturing/freezing. Limited lifespan in vitro, therefore a master bank should be frozen as soon as possible, of at least 10 ampoules.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service