Skip to Content
Merck
All Photos(1)

Key Documents

BCR718

Canned fresh herring

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
BCR718

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: PCB 28, PCB 52, PCB 101, PCB 105, PCB 118, PCB 128, PCB 138, PCB 149, PCB 153, PCB 156, PCB 170, PCB 180

Matrix Group: Fish and Seafood

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service