Skip to Content
Merck
All Photos(1)

Key Documents

MP41G12TR1

Millipore

mPAGE® Trial Kit

includes 2 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MOPS SDS powder, makes 1L of running buffer, 2 tank adapters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NA.56

feature

includes 2 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MOPS SDS powder, makes 1L of running buffer, 2 tank adapters

Looking for similar products? Visit Product Comparison Guide

General description

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

mPAGE 4-12% bis-tris gels for high resolution protein separation
  • Load up to 60 μl sample / well
  • Typical run time 26 min (MES, 180V) or 36 min (MOPS, 200V)
  • Reverse gasket when using with Bio-Rad Mini-PROTEAN systems
  • Use adapter (MPTA) with XCell SureLock and other compatible systems


Western blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.

Protocols

Instructions for setting up an mPAGE SDS-PAGE precast protein gel for gel electrophoresis using a Bio-Rad Mini-PROTEAN® electrophoresis system.

Instructions for setting up an mPAGE SDS-PAGE precast protein gel for gel electrophoresis using a Bio-Rad Mini-PROTEAN® electrophoresis system.

Instructions for setting up an mPAGE SDS-PAGE precast protein gel for gel electrophoresis using a Bio-Rad Mini-PROTEAN® electrophoresis system.

Instructions for setting up an mPAGE SDS-PAGE precast protein gel for gel electrophoresis using a Bio-Rad Mini-PROTEAN® electrophoresis system.

See All

Related Content

mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.

mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.

mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.

mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service