Skip to Content
Merck
All Photos(1)

Key Documents

IM36

Sigma-Aldrich

Anti-MMP-3 (Ab-1) Mouse mAb (55-2A4)

liquid, clone 55-2A4, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

55-2A4, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

porcine, rat, sheep, human

should not react with

bovine, mouse, rabbit

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP3(4314)

General description

Purified mouse monoclonal antibody (see application references). Recognizes the ~59/57 kDa latent and the ~45/28 kDa active forms of MMP-3 and MMP-3/TIMP complexes (see application references).
Recognizes the ~57/59 kDa latent and the ~28/45 kDa active forms of MMP-3. Will also recognize MMP-3/TIMP-1 complexes. Slightly cross-reacts (~34%) with human MMP-10.
This Anti-MMP-3 (Ab-1) Mouse mAb (55-2A4) is validated for use in Frozen Sections, Immunoblotting, Paraffin Sections for the detection of MMP-3 (Ab-1).

Immunogen

Human
human pro-MMP-3 from the conditioned media of rheumatoid synovial fibroblasts

Application


Frozen Sections (see application references)
Immunoblotting (1 g/ml)
Paraffin Sections (see comments and application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Cottam, D.W. and Rees, R.C. 1993. Intl J. Oncol.2, 861.
Stetler-Stevenson, W.G., et al. 1993. FASEB J.7, 1434.
Woessner, J.F. 1991. FASEB J.5, 2145.
Liotta, L.A. and Stetler-Stevenson, W.G. 1990. in Seminars in Cancer Biology, ed. M.M. Gottesman. Vol. 1(2), 99.
This antibody has 34% cross-reactivity with human MMP-10. For paraffin sections, staining can be enhanced by culturing tissue in 2 µM monensin for 3 h. Pre-treating with trypsin or a pressure cooker may enhance staining. Tissue can then be fixed with PLP (perioddate-lysine-4% paraformaldehyde) and stained using 5-10 µg/ml MMP-3 antibody. See application references for more information. Clone 55-2A4 is called Clone B in Obata, K., et al. To prepare conditioned medium for positive control, incubate HT-1080 cells for 2 h at 37°C in serum-free media containing 100 nM PMA. Collect and centrifuge medium; concentrate as necessary. Store in aliquots at -20°C. Antibody should be titrated for optimal results in individual systems.

Legal Information

Manufactured by Daiichi Fine Chemical Co., Ltd. Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bilal Bin Hafeez et al.
Molecular cancer therapeutics, 16(10), 2267-2280 (2017-06-16)
Ormeloxifene is a clinically approved selective estrogen receptor modulator, which has also shown excellent anticancer activity, thus it can be an ideal repurposing pharmacophore. Herein, we report therapeutic effects of ormeloxifene on prostate cancer and elucidate a novel molecular mechanism
Joan Attia-Vigneau et al.
The Journal of investigative dermatology, 134(1), 58-67 (2013-07-03)
Skin aging is characterized by deterioration of the dermal collagen fiber network due to both decreased collagen expression and increased collagenolytic activity. We designed and evaluated in vitro and ex vivo the efficacy of a trifunctional peptide (TFP) to restore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service