Skip to Content
Merck
All Photos(1)

Key Documents

384620

Sigma-Aldrich

Honokiol

A biphenyl neolignan and an active principle of Magnolia extract that displays anti-angiogenic, anti-tumor and anxiolytic properties.

Synonym(s):

Honokiol, 5,3ʹ-Diallyl-2,4ʹ-dihydroxydiphenyl, VEGFR Tyrosine Kinase Inhibitor XIX, VEGFR2 Kinase Inhibitor XV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H18O2/c1-3-5-13-7-9-18(20)16(11-13)14-8-10-17(19)15(12-14)6-4-2/h3-4,7-12,19-20H,1-2,5-6H2

InChI key

FVYXIJYOAGAUQK-UHFFFAOYSA-N

General description

A biphenyl neolignan and an active principle of Magnolia extract that displays anti-angiogenic, anti-tumor and anxiolytic properties. Shown to induce apoptosis and preferentially inhibit the growth of primary human endothelial cells over fibroblasts by blocking VEGFR2 autophosphorylation and subsequent rac activation. Also blocks the in vivo growth of SVR angiosarcoma in nude mice.
The active principle of Magnolia extracts that exhibits anti-angiogenic, anti-tumor, anxiolytic, and chemoprotective properties. Shown to induce apoptosis and preferentially inhibit the growth of primary human endothelial cells over fibroblasts by blocking vascular endothelial growth factor receptor 2 (VEGFR2) autophosphorylation and subsequent rac activation. Also shown to block the growth of SVR angiosarcoma in nude mice.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Anti-angiogenic, anti-tumor and anxiolytic properties
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bai, X., et al. 2003. J. Biol. Chem.278, 35501.
Yang, S.E., et al. 2002. Biochem. Pharmacol.63, 1641.
Son, H.J., et al. 2000. Planta Med.66, 469.
Maruyama, Y., et al. 1998. J. Nat. Prod.61, 135.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service