Skip to Content
Merck
All Photos(1)

Key Documents

344850-M

Millipore

Fumonisin B1, Fusarium moniliforme

A cell-permeable mycotoxin that inhibits sphingolipid biosynthesis in rat kidney and in liver microsomes by inhibition of sphingosine N-acyltransferase (ceramide synthase; IC₅₀ = 100 nM).

Synonym(s):

Fumonisin B1, Fusarium moniliforme, FB₁

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H59NO15
CAS Number:
Molecular Weight:
721.83
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (TLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to beige

solubility

methanol: 5 mg/mL

storage temp.

2-8°C

InChI

1S/C34H59NO15/c1-5-6-9-20(3)32(50-31(44)17-23(34(47)48)15-29(41)42)27(49-30(43)16-22(33(45)46)14-28(39)40)13-19(2)12-24(36)10-7-8-11-25(37)18-26(38)21(4)35/h19-27,32,36-38H,5-18,35H2,1-4H3,(H,39,40)(H,41,42)(H,45,46)(H,47,48)/t19-,20+,21+,22+,23+,24+,25-,26+,27-,32-/m1/s1

InChI key

UVBUBMSSQKOIBE-ZWKVXHQASA-N

General description

A cell-permeable mycotoxin that inhibits sphingolipid biosynthesis in rat kidney and in liver microsomes by inhibition of sphingosine N-acyltransferase (ceramide synthase; IC50 = 100 nM). Preferentially inhibits sphingomyelin biosynthesis in neuronal cells. Exhibits carcinogenic properties.
Fumonisin B1 is a neurotoxin and a phytotoxin. A cell-permeable mycotoxin that inhibits sphingolipid biosynthesis in rat kidney and in liver microsomes by inhibition of sphingosine N-acyltransferase (ceramide synthase) (IC50 = 100 nM). Cellular effects also appear to be induced by micromolar levels of FB1. Because it inhibits ceramide synthase activity, it elevates cellular levels of sphingoid bases, including sphinganine, resulting in overall inhibition of sphingolipid biosynthesis. Preferentially inhibits sphingomyelin biosynthesis in neuronal cells. Has carcinogenic properties.

Biochem/physiol Actions

Primary Target
ceramide synthase
Target IC50: 100 nM against sphingosine N-acyltransferase (ceramide synthase)

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Wu, W.I., et al. 1995. J. Biol. Chem.270, 13171.
Riley, R.T., et al. 1994. J. Nutr. 124, 594.
Wolf, G. 1994. Nutr. Rev. 52, 246.
Merrill, A.H. Jr., et al. 1993. Adv. Lipid Res.26, 215.
Merrill, Jr., A.H., et al. 1993. J. Biol. Chem.268, 27299.
Voss, K.A., et al. 1993. Nat. Toxins1, 222.
Wang, E., et al. 1991. J. Biol. Chem.266, 14486.
Harrison, L.R., et al. 1990. J. Vet. Diag. Invest. 2, 217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service