Skip to Content
Merck
All Photos(1)

Documents

1.46387

Millipore

Thioglycoll. B. clear 750ml

ready-to-use, bottle volume 750 mL , filling volume

Synonym(s):

Fluid Thioglycollate Medium, FTM, Thioglycollate Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

bottle of
single packed of (750 ml in 1 L bottle with white screw cap and septum (6 bottles per box))

bottle capacity

1000 mL

bottle volume

750 mL
750 mL , filling volume

pH

7.1

application(s)

industrial qc
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

aerobic bacteria
anaerobic bacteria
nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Thioglycoll. B. clear contains thioglycollate and L-Cystine that reduce the redox potential of the culture medium and create an anaerobic atmosphere. These components also inactivate mercury and other heavy metal compounds. The reduced agar content and the presence of a clear gelling agent to offer extra clarity. Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.

Application

Fluid Thioglycolate Medium Clear is designed for the isolation and cultivation of fastidious anaerobic and aerobic microorganisms. It is used for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service