Skip to Content
Merck
All Photos(2)

Key Documents

840062C

Avanti

16:0-22:6 PS

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); PS(16:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H73NO10PNa
CAS Number:
Molecular Weight:
830.01
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840062C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840062C

concentration

10 mg/mL (840062C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C44H74NO10P.Na/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-24-26-28-30-32-34-36-43(47)55-40(38-53-56(50,51)54-39-41(45)44(48)49)37-52-42(46)35-33-31-29-27-25-23-16-14-12-10-8-6-4-2;/h5,7,11,13,17-18,20-21,24,26,30,32,40-41H,3-4,6,8-10,12,14-16,19,22-23,25,27-29,31,33-39,45H2,1-2H3,(H,48,49)(H,50,51);/q;+1/p-1/b7-5-,13-11-,18-17-,21-20-,26-24-,32-30-;/t40-,41+;/m1./s1

InChI key

LONMFPKAKXIJFJ-ROYAEOKWSA-M

General description

16:0-22:6 PS or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-L-serine is a glycerophospholipid having, palmitic acid and docosahexaenic acid attached to the sn-1 and sn-2 positions of the glycerol backbone. The phospholipid in this compound is phosphoserine, a negatively charged lipid.

Application

16:0-22:6 PS or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-L-serine (sodium salt) has been used as a calibration standard for lipidomic analysis.

Packaging

5 mL Clear Glass Sealed Ampule (840062C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ségolène Hescot et al.
Oncotarget, 8(66), 109924-109940 (2018-01-05)
Mitotane (o,p'DDD), the most effective drug in adrenocortical carcinoma, concentrates into the mitochondria and impacts mitochondrial functions. To address the molecular mechanisms of mitotane action and to identify its potential target, metabolomic and lipidomic approaches as well as imaging analyses
Marco M Manni et al.
eLife, 7 (2018-03-16)
Phospholipid membranes form cellular barriers but need to be flexible enough to divide by fission. Phospholipids generally contain a saturated fatty acid (FA) at position sn1 whereas the sn2-FA is saturated, monounsaturated or polyunsaturated. Our understanding of the impact of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service