Key Documents
TEM-CF200HCU
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 200 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON 200 HEX COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.