Skip to Content
Merck
All Photos(1)

Key Documents

DWKCPB1000P

Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations

class B, capacity 1000 mL , with +/10mL tolerance

Synonym(s):

CPB1000P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121805
NACRES:
NA.54

feature

class B
graduated

packaging

pkg of 1 ea

manufacturer/tradename

Azlon CPB1000P

capacity

1000 mL , with +/10mL tolerance

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the measurement and dispensing of liquids in the laboratory

Features and Benefits

Conform to ISO 6706 with anti-suction designto prevent base sticking to wet surface

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service