Merck
HomeChemical Analysis for Food and Beverage TestingQuantification of Methylglyoxal in Manuka Honey - A Simple HPTLC Based Approach
Quantification of Methylglyoxal in Manuka Honey - A Simple HPTLC Based Approach
Markus Burholt, Scientist, Instrumental Analytics R&D, Michaela Oberle, Technical Project Manager, Instrumental Analytics R&D, Michael Schulz, Head of HPLC & Instrumental Analytics R&D, Monika Bäumle, Global Product Manager Thin-Layer Chromatography
Merck
Article from Analytix Reporter - Issue 11
Introduction
Honey - a natural product, is one of the most frequently tested food products. In recent years, manuka honey has gained popularity because of its high antibacterial activity.1 Methylglyoxal (MGO) has been identified as one of the major contributors to its antibacterial activity. MGO is present in high concentrations in manuka honey and is directly responsible for its potency. This makes the manuka honey exclusive and high-priced as compared to the other traditional kinds of honey. Manuka honey from New Zealand usually contains 40 to 800 mg/kg of MGO but can even contain up to 1900 mg/kg.2 To avoid adulteration of manuka honey products, a strict quality regulation regarding its origin, purity, and quality needs to be fulfilled and is a prerequisite for the UMF™ (Unique Manuka Factor) grading.2 It mostly reflects the MGO amount in the honey but also considers other authenticity markers.
In the following application, we focus on the MGO quantification using High-Performance Thin-Layer Chromatography (HPTLC) with subsequent substance confirmation by MS measurement. The high viscosity and high sugar content of honey makes it a very complex and matrix-rich sample for an analysis. Thin-Layer Chromatography (TLC) and High-Performance Thin-Layer chromatography (HPTLC) are convenient, fast, and efficient separation techniques that enable the development of analytical methods without the need for complicated sample preparations or high investments.3 Low cost and short analysis time per sample are given by the parallel analysis of many samples on one plate. Furthermore, the high matrix tolerance of TLC offers additional opportunities to existing routine methods.
Experiment: Materials and Methods
Six different commercially available manuka honey samples were analyzed. MGO shows a mesomeric effect and reacts immediately with water to form either methylglyoxal monohydrate or methylglyoxal dihydrate in aqueous environments.4 Only a small amount of around 1% MGO remains unreacted. Direct detection of MGO in manuka honey is found to be difficult using conventional methods. In this application, MGO is converted to stable 2-methylquinoxaline by derivatizing it with 1,2-phenylenediamine (see Figure 1).5
Reaction scheme of Methylglyoxal (MGO) with 1,2-phenylendiamine
Figure 1.Reaction scheme of MGO with 1,2-phenylendiamine
The stable form is then used as the reference. For confirmation of the method and determination of the recovery rate regular honey samples have been spiked with MGO and 1,2-phenylenediamine. Other derivatization options were tested but the reaction with 1,2-phenylenediamine performed best. Water and honey matrix were tested to confirm, that the optimized reaction conditions provide reproducible results for both matrices.
Experimental Conditions
Plate:HPTLC Silica Gel 60 F254 20 x 10 cm (1.05642)
Application volume:0.3 – 9.0 µL, area application 5 x 3 mm with CAMAG ATS 4
Detection:480 nm
Chamber:20x10 chamber without filter paper
Mobile Phase:Ethylacetate/acetonitrile 85:15 (v/v) 
Staining:Anisaldehyde-sulfuric acid reagent (0.5 mL p-anisaldehyde, 85 mL methanol, 10 mL glacial acetic acid, 5 mL sulfuric acid 98%
Migration distance:5 cm
hRf:80
Drying:60 °C
Standard preparation:100 µL of ~40 % aq. methylglyoxal solution (exact content known) diluted in a 20.0 mL volumetric flask and filled up with water. 800 µL of this stock solution is diluted again in a 10.0 mL volumetric flask and made up to the mark with water. Addition of 0.2 % (20 mg) of the reactant 1,2-phenylenediamine. Before the standard was ready for use it was refrigerated at 8 °C for two days to complete the derivatization reaction.
Sample:Solutions of 100 mg/mL of sample nos. 1, 3, 5, and 150 mg/mL of sample nos. 2, 4, 6 were prepared. To every sample 0.2 % of 1,2-phenylenediamine was added. e.g., sample 1: 4.0 g honey diluted in 40 mL solution of water/ethanol in 6:4. To the solution 0.2 % (80 mg) of the reactant 1,2-phenylenediamine was added. Before the samples were ready to use, they were refrigerated at 8 °C for two days to complete the derivatization reaction.
MS measurement:The samples are extracted with the Plate Express and measured with the single-quadrupole expression compact mass spectrometer (CMS) from Advion.
Extraction solvent:Acetonitrile/water 95:5 (v/v) + 0.1% formic acid
A calibration curve of 2-methylquinoxaline was calculated based on 3 different standard volumes (Table 1 and Figure 2).
SpotsApplication volume µLAmount (μg)Mean Area
1, 10, 190.30.0454080.52
2, 11, 201.50.22511120.91
3, 12, 213.00.45115677.39
Table 1.Calibration curve
Calibration curve and calibration function obtained for 2-methylquinoxaline (MGO)
Figure 2.Calibration plot with corresponding calibration function
A recovery study was performed using regular honey to simulate honey matrix. It was spiked with a known amount of MGO standard solution, followed by the addition of 1,2-phenylenediamine. The measured (and calculated) MGO amount allowed for the correlation of the actual amount of MGO in the Manuka honey samples. The experimental details of the recovery rate study can be found in Table 2, Table 3, Figure 3, and Figure 4.
SpotsApplication volume µLDescription
11.0Methylglyoxal standard 0.15 mg/mL (water) with 0.2% 1,2-diphenylenediamine
25.0Regular honey, 100 mg/mL in water/ethanol 3:2
35.0Regular honey, 100 mg/mL in water/ethanol 3:2 + 0.2% 1,2-phenylendiamine
4 - 105.0Regular honey, 100 mg/ml in water/ethanol 3:2 + 0.2% 1,2-phenylendiamine spiked with methylglyoxal 0.024 mg/mL
Table 2.TLC data of recovery rate: In total, nine regular honey samples were applied and one MGO standard sample. Seven honey sample (4-10) were spiked with MGO and 1,2-phenylenediamine.
MeasurementArea
AU
16759.37
26665.00
36911.29
46756.10
57055.36
67059.58
77014.80
Mean Area6888.79
RSD %2.35
Amount (µg)0.108
Spiked Amount (µg)0.12
Recovery rate (%)90.05
Table 3.Quantification of methylglyoxal in the seven honey samples
Visualization of the HPTLC Silica gel 60 F254 plate under visible light (white light); a) matrix compounds after staining with anisaldehyde sulfuric acid (black areas); b) 2-Methylquinoxaline (blue spot at hRf 80), (reaction product of Methylglyoxal with 1,2-phenylenediamine)
Figure 3.Visualization of the plate under visible light (white light); a) matrix compounds after staining with anisaldehyde sulfuric acid (blackareas); b) 2-Methylquinoxaline (blue spot at hRf 80), (reaction product of Methylglyoxal with 1,2-phenylenediamine)
Scan of spiked honey tracks (sample 4 – 10) at 480 nm with CAMAG TLC Scanner 3
Figure 4.Scan of spiked honey tracks (sample 4 – 10) at 480 nm with CAMAG TLC Scanner 3
All TLC analyses were performed using HPTLC Silica gel 60 F254. The plates were pre-washed with the mobile phase (up to 7 cm) and dried before use.
The standards were prepared by dissolving 100 µL of ~40% aq. MGO solution (exact content known) diluted in 20.0 mL water. 800 μL of this stock solution was further diluted with water to 10.00 mL volume and 0.2% (20 mg) of the reactant 1,2-phenylenediamine was added. All standard solutions were stored at 8 °C for two days before use to achieve reproducible reaction of MGO with 1,2-phenylenediamine. Longer storage times (>3 days) lead to partly degradation of 2-methylquinoxaline.
Honey sample solutions of 100 mg/mL in case of sample numbers 1, 3, 5, and 150 mg/mL in case of honey samples 2, 4 and 6 were applied with a higher volume due to the expected lower amount of MGO. To each sample 0.2% of 1,2-phenylenediamine was added, e.g., sample 1, 4.0 g honey diluted in 40 mL solution of water / ethanol in 3:2. To the solution 0.2% (80 mg) of the reactant 1,2-phenylenediamine was added. Before using the samples, they needed to be stored at 8 °C for two days to complete the reaction.
The samples and standards were applied as spots in an area of 5 x 3 mm.2  This step is necessary because of the high matrix and high application volumes of the honey samples. The plate was developed, dried, and then derivatized by dipping in an anisaldehyde-sulfuric acid reagent. Blue spots of 2-methylquinoxaline (product of the reaction of MGO with 1,2‑phenylenediamine) appeared at hRf 80. Daylight examination and scanning of the plate at 480 nm were carried out for quantification. Experimental results are shown in Figure 5 and Table 4.
Visualization of the HPTLC gel 60 F254 plate under visible light (white light); a) matrix compounds after staining with anisaldehyde sulfuric acid (black areas);b) 2-methylquinoxaline (blue spot at hRf 80), (reaction product of methylglyoxal with 1,2-phenylenediamine)
Figure 5.Visualization of the plate under visible light (white light); a) matrix compounds after staining with anisaldehyde sulfuric acid (black areas); b) 2-methylquinoxaline (blue spot at hRf 80), (reaction product of methylglyoxal with 1,2-phenylenediamine)
  Manuka Samples  Application positionConc. Sample (mg/mL)Application volume (µL)  Mean Area (AU)Mean Amount (µg)    %RSD  MGO in Honey (mg/kg)Expected Amount MGO in Honey according to information on product label (mg/kg)
14, 13, 22100.05.011225.000.2282.68507.4600.0
25, 14, 23150.09.04548.800.0552.8445.3nd
36, 15, 24100.05.08002.840.1363.10301.4300.0
47, 16, 25150.08.06031.580.0882.4881.480.0
58, 17, 26100.05.09674.570.1813.06401.8400.0
69, 18, 27150.08.07578.220.1253.14115.6nd
Table 4.TLC data: In total 27 samples were applied. Track numbers with applied samples and volumes and obtained hRf values are summarized here (details of tracks 1-3, 10-12 and 19-21 for calibration are given in Table 1).
A separate plate without staining was used for MS measurement. The coupling to MS was performed on an elution-based approach, that utilized a TLC-MS interface. This enabled the dissolution of the analyte from the silica plate at the zone of hRf 80 by a solvent and a transfer to the mass spectrometer in the liquid phase. This additionally confirmed the spot identification of the MGO derivative 1-methylquinoxaline.6
Results and Discussion
As demonstrated, MGO can be identified and quantified in different honey samples within the concentration range of 50 mg to 600 mg/kg. The conversion of MGO into the more stable compound 2-methylquinoxaline allows for an easy evaluation of the MGO content. The recovery study showed a detectable MGO amount of around 90%. The correlated MGO amount in manuka samples was calculated accordingly. One of the samples (sample 1) showed a lower MGO content than indicated by the supplier. This might be because of the degradation of the MGO during storage. Sample 2 and sample 6 only showed MGO concentrations of 50 and 100 mg/kg. These manuka honey samples are considered of lower quality. Although no indication of MGO concentration was provided by the supplier.
Conclusion
The analysis of MGO in a complex and challenging food matrix like honey was described. Target analyte could be easily separated and detected without time-consuming and labor-intensive sample preparation. The flexible instrument set-up enabled a combination with the MS measurements.
Screening and method development capabilities were shown by the application of 27 tracks in one plate (honey samples and standard solutions). The study clearly emphasized different honey qualities (referring to MGO content) on the market. Though the analysis of MGO is challenging, MGO content could be identified in the expected range.
To summarize, a fast, cheap, and simple quantification of methylglyoxal can be accomplished with HPTLC. This application demonstrates the main advantages of the method, such as quick sample preparation, high-matrix tolerance, and high-throughput.
Related Application Categories
Chemical Analysis for Food and Beverage Testing
Thin Layer Chromatography
See also information about the TLC Explorer Documentation System,  a superior instrumental and software solution for reliable TLC plate analysis.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
TLC-MS
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Mavric E, Wittmann S, Barth G, Henle T. Identification and quantification of methylglyoxal as the dominant antibacterial constituent of Manuka (Leptospermum scoparium)honeys from New Zealand. Mol. Nutr. Food Res.. 2008;52(4):483-489. https://doi.org/10.1002/mnfr.200700282
2.
Manuka Honey MGO vs UMF™ Calculator. [Internet]. Available from: https://newzealandhoneyco.com/pages/mgo-vs-umf-calculator-manuka-honey
3.
Schulz M, Burhold M, Baeumle M. 50 years of TLC-MS, Analytix Reporter 2019;5;3-7. [Internet]. Available from: https://www.sigmaaldrich. com/deepweb/assets/sigmaaldrich/marketing/global/ documents/730/477/analytix-reporter-5-2019-mk.pdf#page=3
4.
Identification and quantification of methylglyoxal in Manuka honey, magritek. [Internet]. (Accessed Nov. 2021). Available from: https://magritek.com/2021/01/29/identification-and-quantification-of-methylglyoxal-in-manuka-honey/
5.
E. Dimitrova, Bestimmung von Glyoxal und Methylglyoxal in Lebensmitteln und im Serum exponierter Mäuse - Eine Basis für die Abschätzung der alimentären Exposition und Bioverfügbarkeit, 2013, Master thesis, TU University of Graz, Austria. [Internet]. (Accessed Nov. 2021). Available from: https://diglib.tugraz.at/download.php?id=576a76ef4c784&location=browse
6.
unpublished results.