Merck
HomeSmall Molecule HPLCEvaluation of Different Enzymes on Hydrolysis Efficiencies of Glucuronide Drug Metabolites in Urine
Evaluation of Different Enzymes on Hydrolysis Efficiencies of Glucuronide Drug Metabolites in Urine
Jim Blasberg, Harkewal Singh, Kevin Ray
Sigma-Aldrich, St. Louis, MO
Introduction
β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS. Here we evaluate the hydrolysis efficiency of β-glucuronidase derived from Patella vulgata (limpet), Helix pomatia (snail), Red abalone, and E. coli on drugs of abuse and pain management in a synthetic urine matrix. Hydrolysis efficiencies vary from product to product and several parameters need to be investigated in determining which enzyme to use.
Background
dimer
Dimer of β-GUS (inset: expanded view of active site)
Common Currency for Glucuronidase Activity
typical-reaction
One Fishman1 unit will liberate 1 μg phenolphthalein from phenolphthalein glucuronide per hour at 37οC All hydrolysis experiments were normalized for enzyme activity, i.e. equivalent Fishman units
Methods
  • Target glucuronides were spiked into synthetic urine.
  • Heavy parent (non-glucuronide) drug was spiked as an internal standard.
  • Added enzyme at an appropriate pH, incubated at 60 °C.
  • Each time-point pulled sample and precipitate protein with TCA.
  • Analyzed by LC-MS/MS (typical conditions below).
methods
Results
codeine-glucuronide
Codeine-gluc enzyme source
  • Enzyme added at 10 units/μL of urine and incubated at 60 °C.
  • Codeine glucuronide conversion was enzyme source dependent at 10 U/μL, 60 °C.
  • E. coli enzyme shows high activity early but appears to lose activity after 1 hour at 60 °C.
  • Limpet enzyme shows >95% codeine glucuronide conversion at 10 U/μL, 60 °C.
temazepam-glucuronide
Temazepam-gluc enzyme source
  • Enzyme added at 1 unit/μL of urine and incubated at 60 °C.
  • Temazepam glucuronide conversion was fast at 1 U/μL, 60 °C, for all enzymes tested.
  • Most benzodiazepine glucuronides show full conversion even at low titer and 0.5 hr @ 60C
Purified Abalone GUS
purified-abalone-gus
  • Oxymorphone glucuronide hydrolysis was equivalent at 10 U/μL, 60 °C, for purified abalone GUS and recombinant GUS.
Recombinant GUS Comparison - 6MAM Conversion
recombinant-gus-comparison
  • Traditional abalone exhibited substantial 6-MAM to morphine conversion.
  • 6-MAM conversion at 75 U/μL, 60 °C, was equivalent for purified abalone GUS and recombinant GUS.
Effect of Enzyme Titer - Purified Abalone
codeine-glucuronide-2
Codeine-gluc enzyme titer
  • Purified abalone enzyme added at 1, 10, and 75 units/μL of urine and incubated at 60 °C.
  • Enzyme titer greatly affects conversion rate
  • Hydrolysis of codeine glucuronide completes in less than an hour at 75 U/μL.
temazepam-glucuronide-2
Temazepam-gluc enzyme titer
  • Purified abalone enzyme added at 0.1, 1, and 10 units/μL of urine and incubated at 60 °C
  • Temazepam glucuronide conversion was less dependent on enzyme titer.
  • Hydrolysis of temazepam glucuronide completes in less than an hour at 1 U/μL.
Conclusions
  • Effect of Enzyme Source
    - Glucuronide conversion efficiency is enzyme-dependent.
    - Glucuronide conversion efficiency is also substrate-dependent.
  • Effect of Enzyme Titer
    - At high titer even difficult to convert substrates, such as codeine, are hydrolyzed rapidly.
    - In general benzodiazepines, such as temazepam, hydrolyze rapidly even at low titers.
  • Comparison of Purified Abalone and Recombinant GUS
    - Purified abalone hydrolyzed oxymorphone glucuronide quickly at 10 U/μL, comparable to recombinant GUS.
    - Purified abalone induced minimal 6-monoacetyl morphine (6-MAM) conversion even at high titer.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Reference
1.
Fishman WH. 1974. ?-Glucuronidase.929-943. https://doi.org/10.1016/b978-0-12-091302-2.50082-7