MilliSentials Lab Labeling System Setup
MilliSentials™ Lab Labeling System
The MilliSentials™ Lab Labeling System has been designed as a complete solution for laboratory labeling. As part of that solution we’ve put together a how-to-setup video, troubleshooting section, and list of materials to support the system.
How to Setup the MilliSentials™ Lab Labeling System
The following video provides a complete step by step procedure to unbox, setup, and use the MilliSentials™ Lab Labeling System. The video is composed of eight sections which can be directly accessed within the video by following the links included below.
Video TimeStamps
- (0:00): Section 1 - Introduction
- (0:21): Section 2 - Contents & Unpacking Your System
- (0:55): Section 3 - Device Requirements & Installation of Required Software
- (2:40): Section 4 - Installation of MilliSentials™ Lab Ribbon
- (5:13): Section 5 - Installation of MilliSentials™ Lab Adapatable Labels
- (6:10): Section 6 - Deciding Your Printer Connection & Set Up Preferences
- (8:32): Section 7 - Connecting Your Printer via USB Cable with a Windows Computer
- (10:59): Section 8 - Connecting Your Printer to the Network via WiFI
- (14:30): Section 9 - Using the MilliSentials™ Lab Labeling System Software
- (20:25): Section 10 - Using the MilliSentials™ Lab Labeling System to Print
Millisentials™ Lab Labeling System Barcoding Software Demonstration
View this video for a complete tutorial on how to create labels using the intuitive system software. Includes step-by-step instructions on creating 2D and 3D barcodes for scannable labels.
Troubleshooting | |
---|---|
Question | Proposed Solution(s) |
During installation, my printer did not re-start when I ran through my set up process. |
|
I tried printing and the text is light, spotty, or incomplete. | Reset the printer settings. Plug in the Flash Drive included with the system and navigate to the folder using file explorer. Double click on the printer settings. Reset the printer. Retry printing.
|
I tried printing but the text came out misaligned. | Reset the printer settings. Plug in the Flash Drive included with the system and navigate to the folder using file explorer. Double click on the printer settings. Reset the printer. Retry printing.
|
I tried printing and no text was printed. |
|
I tried printing labels, but the label roller won’t move and is not even producing blank labels. |
|
When setting up my wireless printer, it keeps asking for a password but I’m using an open network. | Since you are using an open network, there are less security options. While setting up in Zebra Setup Utilities, under security, click “none”. Then in the next screen select “hex” and do not enter any passwords.
If the problem persists and the printer will not print, contact your IT administrator. |
After I closed my printer, it rejected a large number of labels and then concluded with MEDIA OUT error message. How do I fix this? |
|
Why does the printer reject three or four labels every time I turn the printer on, press the pause/un-pause button or open the printer head? | The printer is using self-calibration mode. If you wish to remove this functionality, you may go to the printer hardware, from the main menu, click TOOLS → Power up action and set it to “no motion”. Then, under TOOLS → Head close action, set it to “no motion”. Just make sure that when you select this, you press the “feed button” (center, next to the pause button) once or twice to correctly align your labels before printing after opening the printer head.
|
I can’t find Zebra Setup Utilities or can’t install the Zebra Setup Utilities program on my Mac device. | There is no Zebra Setup Utilities program designed for Mac. Refer to our video or the User Guide for how to connect your Mac device to the printer and MilliSentials™ Lab Labeling System Software
|
Does the printer always have to be connected to a laptop in the lab? | No. Refer to wireless connection instructions in the full user guide or how-to video. You may also use your mobile device to print labels.
|
I went through the steps to setup the printer on the ethernet or WIFI network, but the printer icon is still red. |
|
Materials | |||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Name | Description | Country/Region | SKU | ||||||||||||||||||||
MilliSentials™ Lab Labeling System | Labeling System including the following items:
|
| |||||||||||||||||||||
MilliSentials™ Lab Labels | MilliSentials™ Lab Adaptable Labels - 1 roll of labels that contain 1615 labels | Global | MISELADLA | ||||||||||||||||||||
MilliSentials™ Lab Ribbon | MilliSentials™ Lab Ribbon – 1 ribbon that provides printing for 6000 labels | Global | MISELPRRI |