Sample Preparation for Chromatographic Purification
Samples for chromatographic purification should be clear and free from particulate matter. Simple steps to clarify a sample before beginning purification will avoid clogging the column, may reduce the need for stringent washing procedures, and can extend the life of the chromatographic medium.
Sample extraction procedures and the selection of buffers, additives, and detergents are determined largely by the source of the material, the stability of the target molecule, the chromatographic techniques that will be employed, and the intended use of the product. These subjects are dealt with in general terms in the Protein Purification Handbook and more specifically according to target molecule in the Recombinant Protein Handbook, Protein Amplification and Simple Purification, and Antibody Purification Handbook, available from Cytiva.
Sample stability
In the majority of cases, biological activity needs to be retained after purification. Retaining the activity of the target molecule is also an advantage when following the progress of the purification, since detection of the target molecule often relies on its biological activity. Denaturation of sample components often leads to precipitation or enhanced non-specific adsorption, both of which will impair column function. Hence, there are many advantages to checking the stability limits of the sample and working within these limits during purification.
Proteins generally contain a high degree of tertiary structure, kept together by van der Waals' forces, ionic and hydrophobic interactions, and hydrogen bonding. Any conditions capable of destabilizing these forces may cause denaturation and/or precipitation. By contrast, peptides contain a low degree of tertiary structure. Their native state is dominated by secondary structures, stabilized mainly by hydrogen bonding. For this reason, peptides tolerate a much wider range of conditions than proteins. This basic difference in native structures is also reflected in that proteins are not easily renatured, while peptides often renature spontaneously.
It is advisable to perform stability tests before beginning to develop a purification protocol. The list below may be used as a basis for such testing:
- Test pH stability in steps of one pH unit between pH 2 and pH 9.
- Test salt stability with 0–2 M NaCl and 0–2 M (NH4)2SO4 in steps of 0.5 M.
- Test the stability toward acetonitrile and methanol in 10% steps between 0% and 50%.
- Test the temperature stability in 10 °C steps from 4 °C to 40 °C.
- Test the stability and occurrence of proteolytic activity by leaving an aliquot of the sample at room temperature overnight. Centrifuge each sample and measure activity and UV absorbance at 280 nm in the supernatant.
Sample clarification
Centrifugation and filtration are standard laboratory techniques for sample clarification and are used routinely when handling small samples.
Centrifugation and filtration are standard laboratory techniques for sample clarification and are used routinely when handling small samples.
It is highly recommended to centrifuge and filter any sample immediately before chromatographic purification.
Centrifugation
Centrifugation removes lipids and particulate matter, such as cell debris. If the sample is still not clear after centrifugation, use filter paper or a 5 μm filter as a first step and one of the filters below as a second step filter.
- For small sample volumes or proteins that adsorb to filters, centrifuge at 10 000 ×g for 15 min.
- For cell lysates, centrifuge at 40 000–50 000 ×g for 30 min.
- Serum samples can be filtered through glass wool after centrifugation to remove any remaining lipids.
Filtration
Filtration removes particulate matter. Membrane filters that give the least amount of non-specific binding of proteins are composed of cellulose acetate or PVDF.
For sample preparation before chromatography, select a filter pore size in relation to the bead size of the chromatographic medium.
Nominal pore size of filter | Particle size of chromatographic medium |
---|---|
1 μm | 90 μm and upward |
0.45 μm | 30 or 34 μm |
0.22 μm | 3, 10, 15 μm or when extra clean samples or sterile filtration is required |
Check the recovery of the target protein in a test run. Some proteins may adsorb non-specifically to filter surfaces.
Desalting
Desalting columns are suitable for any sample volume and will rapidly remove low molecular weight contaminants in a single step at the same time as transferring the sample into the correct buffer conditions. Centrifugation and/or filtration of the sample before desalting is still recommended. Detailed procedures for buffer exchange and desalting are given on page 136 [BS1].
At laboratory scale, when samples are reasonably clean after filtration or centrifugation, the buffer exchange and desalting step can be avoided. For affinity chromatography or hydrophobic interaction chromatography, it may be sufficient to adjust the pH of the sample and, if necessary, dilute to reduce the ionic strength of the solution.
- Rapidly process small or large sample volumes. Use before and/or between purification steps, if needed (remember that each extra step can reduce yield and desalting also dilutes the sample).
- Remove salts from proteins with molecular weight Mr > 5000.
- Use 100 mM ammonium acetate or 100 mM ammonium hydrogen carbonate if volatile buffers are required.
Specific sample preparation steps
Specific sample preparation steps may be required if the crude sample is known to contain contamininants such as lipids, lipoproteins, or phenol red that may build up on a column or if certain gross impurities, such as bulk protein, should be removed before any chromatographic step.
Fractional precipitation
Fractional precipitation is frequently used at laboratory scale to remove gross impurities from small sample volumes, and occasionally used in small-scale commercial production. Precipitation techniques separate fractions by the principle of differential solubility. Because protein species differ in their degree of hydrophobicity, increased salt concentrations can enhance hydrophobic interactions between the proteins and cause precipitation. Fractional precipitation can be applied to remove gross impurities in three different ways, as shown in Figure 86.
Figure 86.Three ways to use precipitation.
Examples of precipitation agents are reviewed in Table 27. The most common precipitation method using ammonium sulfate is described in more detail.
Precipitation agent | Typical conditions for use | Sample type | Comment |
---|---|---|---|
Ammonium sulfate | As described below. | > 1 mg/mL proteins especially immuno-globulins. | Stabilizes proteins, no denaturation; supernatant can go directly to HIC. Helps to reduce lipid content. |
Dextran sulfate | Add 0.04 mL 10% dextran sulfate and 1 mL 1 M CaCl2 per mL sample, mix 15 min, centrifuge 10 000 ×g, discard pellet. | Samples with high levels of lipoprotein e.g., ascites. | Precipitates lipoprotein. |
Polyvinylpyrrolidine | Add 3% (w/v), stir 4 h, centrifuge 17 000 ×g, discard pellet. | Samples with high levels of lipoprotein e.g., ascites. | Alternative to dextran sulfate. |
Polyethylene glycol (PEG, Mr > 4000) | Up to 20% w/vol | Plasma proteins. | No denaturation, supernatant goes directly to IEX or AC, complete removal may be difficult. Stabilizes proteins. |
Acetone (cold) | Up to 80% vol/ at 0°C. Collect pellet after centrifugation at full speed in Eppendorf™ centrifuge. | May denature protein irreversibly. Useful for peptide precipitation or concentration of sample for electrophoresis. | |
Polyethyleneimine | 0.1% w/v | Precipitates aggregated nucleoproteins. | |
Protamine sulfate | 1% w/v | Precipitates aggregated nucleoproteins. | |
Streptomycin sulfate | 1% w/v | Precipitation of nucleic acids. | |
Caprylic acid | X/15) g where X = volume of sample. | Antibody concentration should be > 1 mg/mL. | Precipitates bulk of proteins from sera or ascites, leaving immunoglobulins in solution. |
Details taken from: Scopes R.K., Protein Purification, Principles and Practice, Springer, (1994), J.C. Janson and L. Rydén, Protein Purification, Principles, High Resolution Methods and Applications, 2nd ed. Wiley Inc, (1998).
Personal communications.
Ammonium sulfate precipitation
Some proteins may be damaged by ammonium sulfate. Take care when adding crystalline ammonium sulfate: high local concentrations may cause contamination of the precipitate with unwanted proteins.
For routine, reproducible purification, precipitation with ammonium sulfate should be avoided in favor of chromatography.
In general, precipitation is rarely effective for protein concentrations below 1 mg/mL.
Solutions needed for precipitation:
Saturated ammonium sulfate solution (add 100 g ammonium sulfate to 100 mL distilled water, stir to dissolve).
1 M Tris-HCl, pH 8.0.
Buffer for first purification step
- Filter (0.45 μm) or centrifuge the sample (10 000 ×g at 4 °C).
- Add 1 part 1 M Tris-HCl, pH 8.0 to 10 parts sample volume to maintain pH.
- Stir gently. Add ammonium sulfate solution, drop by drop. Add up to 50% saturation*. Stir for 1 hour.
- Centrifuge 20 min at 10 000 ×g.
- Remove supernatant. Wash the pellet twice by resuspension in an equal volume of ammonium sulfate solution of the same concentration (i.e., a solution that will not redissolve the precipitated protein or cause further precipitation). Centrifuge again.
- Dissolve pellet in a small volume of the buffer to be used for the next step.
- Ammonium sulfate is removed during clarification/buffer exchange steps with Sephadex™ G-25, using desalting columns (see page 136).
*The % saturation can be adjusted either to precipitate a target molecule or to precipitate contaminants.
The quantity of ammonium sulfate required to reach a given degree of saturation varies according to temperature. Table 28 shows the quantities required at 20 °C.
Final percent saturation to be obtained+A1:R23 | |||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
20 | 25 | 30 | 35 | 40 | 45 | 50 | 55 | 60 | 65 | 70 | 75 | 80 | 85 | 90 | 95 | 100 | |
Starting percent saturation | Amount of ammonium sulfate to add (grams) per liter of solution at 20 °C | ||||||||||||||||
0 | 113 | 144 | 176 | 208 | 242 | 277 | 314 | 351 | 390 | 430 | 472 | 516 | 561 | 608 | 657 | 708 | 761 |
5 | 85 | 115 | 146 | 179 | 212 | 246 | 282 | 319 | 358 | 397 | 439 | 481 | 526 | 572 | 621 | 671 | 723 |
10 | 57 | 86 | 117 | 149 | 182 | 216 | 251 | 287 | 325 | 364 | 405 | 447 | 491 | 537 | 584 | 634 | 685 |
15 | 28 | 58 | 88 | 119 | 151 | 185 | 219 | 255 | 293 | 331 | 371 | 413 | 456 | 501 | 548 | 596 | 647 |
20 | 0 | 29 | 59 | 89 | 121 | 154 | 188 | 223 | 260 | 298 | 337 | 378 | 421 | 465 | 511 | 559 | 609 |
25 | 0 | 29 | 60 | 91 | 123 | 157 | 191 | 228 | 265 | 304 | 344 | 386 | 429 | 475 | 522 | 571 | |
30 | 0 | 30 | 61 | 92 | 125 | 160 | 195 | 232 | 270 | 309 | 351 | 393 | 438 | 485 | 533 | ||
35 | 0 | 30 | 62 | 94 | 128 | 163 | 199 | 236 | 275 | 316 | 358 | 402 | 447 | 495 | |||
40 | 0 | 31 | 63 | 96 | 130 | 166 | 202 | 241 | 281 | 322 | 365 | 410 | 457 | ||||
45 | 0 | 31 | 64 | 98 | 132 | 169 | 206 | 245 | 286 | 329 | 373 | 419 | |||||
50 | 0 | 32 | 65 | 99 | 135 | 172 | 210 | 250 | 292 | 335 | 381 | ||||||
55 | 0 | 33 | 66 | 101 | 138 | 175 | 215 | 256 | 298 | 343 | |||||||
60 | 0 | 33 | 67 | 103 | 140 | 179 | 219 | 261 | 305 | ||||||||
65 | 0 | 34 | 69 | 105 | 143 | 183 | 224 | 267 | |||||||||
70 | 0 | 34 | 70 | 107 | 146 | 186 | 228 | ||||||||||
75 | 0 | 35 | 72 | 110 | 149 | 190 | |||||||||||
80 | 0 | 36 | 73 | 112 | 152 | ||||||||||||
85 | 0 | 37 | 75 | 114 | |||||||||||||
90 | 0 | 37 | 76 | ||||||||||||||
95 | 0 | 38 |
Resolubilization of protein precipitates
Many proteins are easily resolubilized in a small amount of the buffer to be used in the next chromatographic step. However, a denaturing agent may be required for less soluble proteins. Specific conditions will depend upon the specific protein. These agents must always be removed to allow complete refolding of the protein and to maximize recovery of mass and activity. A chromatographic step often removes a denaturant during purification. Table 29 gives examples of common denaturing agents.
Denaturing agent | Typical conditions for use | Removal/comment |
---|---|---|
Urea | 2 M–8 M | Remove using Sephadex G-25. |
Guanidine hydrochloride | 3 M–6 M | Remove using Sephadex G-25 or during IEX. |
Triton X-100 | 2% | Remove using Sephadex G-25 or during IEX. |
Sarcosyl | 1.5% | Remove using Sephadex G-25 or during IEX. |
N-octyl glucoside | 2% | Remove using Sephadex G-25 or during IEX. |
Sodium dodecyl sulfate | 0.1%–0.5% | Exchange for non-ionic detergent during first chromatographic step, avoid anion exchange chromatography. |
Alkaline pH | > pH 9, NaOH | May need to adjust pH during chromatography to maintain solubility. |
Details taken from: Scopes R.K., Protein Purification, Principles and Practice, Springer, (1994), J.C. Janson and L. Rydén, Protein Purification,
Principles, High Resolution Methods and Applications, 2nd ed. Wiley Inc, (1998) and other sources.
Buffer exchange and desalting
Dialysis is frequently mentioned in the literature as a technique to remove salt or other small molecules and to exchange the buffer composition of a sample. However, dialysis is generally a very slow technique, requiring large volumes of buffer. During handling or as a result of proteolytic breakdown or non-specific binding to the dialysis membranes, there is a risk of losing material. A simpler and much faster technique is to use a desalting column, packed with Sephadex G-25, to perform a group separation between high and low molecular weight substances. Proteins are separated from salts and other small molecules.
In a fast, single step, the sample is desalted, transferred into a new buffer and low molecular weight materials are removed.
Desalting columns are used not only to remove low molecular weight contaminants, such as salt, but also for buffer exchange before or after different chromatographic steps and for the rapid removal of reagents to terminate a reaction.
Sample volumes up to 30% of the total volume of the desalting column can be processed. Sample concentration does not influence the separation as long as the concentration of proteins does not exceed 70 mg/mL when using normal aqueous buffers. The sample should be fully dissolved. Centrifuge or filter to remove particulate material.
For small sample volumes it may be possible to dilute the sample with the start buffer that is to be used for chromatographic purification, but cell debris and particulate matter must still be removed.
To prevent possible ionic interactions the presence of a low salt concentration (25 mM NaCl) is recommended during desalting and in the final sample buffer.
Volatile buffers such as 100 mM ammonium acetate or 100 mM ammonium hydrogen carbonate can be used if it is necessary to avoid the presence of NaCl.
Figure 87.Buffer exchange of mouse plasma (10 mL) on HiPrep™ 26/10 Desalting.
For laboratory-scale operations, Table 30 shows a selection guide for prepacked, ready to use desalting and buffer exchange columns.
Column | Sample volume | Sample elution volume |
---|---|---|
MicroSpin™ G-25 | 0.1–0.15 mL | 0.1–0.15 mL |
PD-10 (gravity column) | 1.5–2.5 mL | 2.5–3.5 mL |
HiTrap™ Desalting 5 mL | 0.25–1.5 mL | 1.0–2.0 mL |
HiPrep™ 26/10 Desalting | 2.5–15 mL | 7.5–20 mL |
To desalt larger sample volumes:
- connect up to 5 HiTrap™ Desalting 5 mL columns in series to increase the sample volume capacity, for example, 2 columns: sample volume 3 mL, 5 columns: sample volume 7.5 mL.
- connect up to 4 HiPrep™ 26/10 Desalting columns in series to increase the sample volume capacity, for example, 2 columns: sample volume 30 mL, 4 columns: sample volume 60 mL. Even with 4 columns in series, the sample can be processed in 20 to 30 min, at room temperature, in aqueous buffers.
Instructions are supplied with each column. Desalting and buffer exchange can take less than 5 min per sample with greater than 95% recovery for most proteins.
Alternative 1: Manual desalting with HiTrap™ Desalting 5 mL using a syringe
- Fill the syringe with buffer. Remove the stop plug. To avoid introducing air into the column, connect the column "drop to drop" to the syringe (via the adapter provided).
- Remove the snap-off end at the column outlet.
- Wash the column with 25 mL buffer at 5 mL/min to remove completely the 20% ethanol (supplied as storage buffer). If air is trapped in the column, wash with degassed buffer until the air disappears. Air bubbles introduced onto the column by accident during sample application do not influence the separation.
- Apply the sample using a 2–5 mL syringe at a flow rate between 1–10 mL/min. Discard the liquid eluted from the column.
- If the sample volume is less than 1.5 mL, change to buffer and proceed with the injection until a total of 1.5 mL has been eluted. Discard the eluted liquid.
- Elute the protein with the appropriate volume selected from Table 31.
Collect the desalted protein in the volume indicated.
Note: 5 mL/min corresponds to approximately 120 drops/min when using a HiTrap™ 5 mL column. A simple peristaltic pump can also be used to apply sample and buffers.
The maximum recommended sample volume is 1.5 mL. See Table 31 for the effect of reducing the sample volume applied to the column.
A simple peristaltic pump can also be used to apply sample and buffers.
Sample load mL | Add buffer mL | Elute and collect mL | Yield % | Remaining salt % | Dilution factor |
---|---|---|---|---|---|
0.25 | 1.25 | 1.0 | >95 | 0.0 | 4.0 |
0.50 | 1.0 | 1.5 | >95 | <0.1 | 3.0 |
1.00 | 0.5 | 2.0 | >95 | <0.2 | 2.0 |
1.50 | 0 | 2.0 | >95 | <0.2 | 1.3 |
Alternative 2: Simple desalting with ÄKTAprime Plus
ÄKTAprime™ Plus contains pre-programmed templates for individual HiTrap™ Desalting 5 mL and HiPrep™ 26/10 Desalting columns.
Buffer Preparation
Prepare at least 500 mL of the required buffer.
- Follow the instructions supplied on the ÄKTAprime Plus cue card to connect the column and load the system with buffer.
- Select the Application Template.
- Start the method.
- Enter the sample volume and press OK.
Figure 88 shows a typical result obtained from ÄKTAprime Plus. The UV (protein) and conductivity (salt) traces enable pooling of the desalted fractions.
Figure 88.Desalting of a (Histidine)6-tagged fusion protein on ÄKTAprime Plus.
Removal of lipoproteins
Lipoproteins and other lipid material can rapidly clog chromatography columns and it is advisable to remove them before beginning purification. Precipitation agents such as dextran sulfate and polyvinylpyrrolidine, described under Fractional precipitation, are recommended to remove high levels of lipoproteins from samples such as ascitic fluid.
Centrifuge samples to avoid the risk of non-specific binding of the target molecule to a filter.
Samples such as serum can be filtered through glass wool to remove remaining lipids.
Removal of phenol red
Phenol red is frequently used at laboratory scale as a pH indicator in cell culture. Although not directly interfering with purification, phenol red may bind to certain purification media and should be removed as early as possible to avoid the risk of contamination. It is known to bind to anion exchange media at pH > 7.
Use a desalting column to simultaneously remove phenol red (a low molecular weight molecule) and transfer sample to the correct buffer conditions for further purification, as described under Buffer exchange and desalting.
Removal of low molecular weight contaminants
If samples contain a high level of low molecular weight contaminants, use a desalting column before the first chromatographic purification step, as described under Buffer exchange and desalting.
Network error: Failed to fetch