Merck
HomeGas Chromatography (GC) EN 14105: GC Analysis of Glycerin Impurity in Biodiesel on MET-Biodiesel
EN 14105: GC Analysis of Glycerin Impurity in Biodiesel on MET-Biodiesel
EN 14105: GC Analysis of Glycerin Impurity in Biodiesel on MET-Biodiesel
Conditions
columnMET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven50 °C (1 min), 15 °C/min to 180 °C, 7 °C/min to 230 °C, 30 °C/min to 380 °C (10 min)
carrier gashelium, 3.0 mL/min
sampleEN 14105:2003 Monoglyceride Stock Solution (49446-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA then diluted in heptane
injection1 µL, cold on-column
detectorFID, 380 °C
Description
CategoriesAnalytical Chromatography, GC Applications, Glycerides
Featured IndustryAgriculture
Biofuels
Other NotesComplementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitabilityapplication for GC