Merck
HomeMILLIPLEX® Multiplex for Luminex® ImmunoassaysMultiplex Analysis: Hepatotoxic Biomarkers
Multiplex Analysis: Hepatotoxic Biomarkers
Read more about
The list of hepatotoxic biomarkers in drug-induced liver injury is constantly expanding. Multiplex assays, such as the Luminex® bead-based multiplex immunoassay MILLIPLEX® Rat Liver Injury Panel, enable researchers to investigate multiple hepatotoxic biomarkers simultaneously, saving time and sample. Multiplexed analysis can also measure cytokine levels and allow for correlations between pro-inflammatory markers and liver injury, furthering the understanding of liver injury processes.
What Are Biomarkers of Drug-Induced Liver Injury?
Drug-induced liver injury (DILI) is the primary adverse event that results in the withdrawal of drugs from the market.1 The liver is a multifunctional organ. As drugs may cause liver injury through various mechanisms, measurement of multiple biomarkers is required to ensure the potential for hepatotoxicity is identified early in drug development.2 Traditionally, the biomarkers of drug-induced liver injury have been alanine aminotransferase (ALT) and aspartate aminotransferase (AST) which are used to assess liver injury. However, with significant frequency, levels of these biomarkers do not accurately reflect injury as seen by histopathology.3 Therefore, the Predictive Safety Testing Consortium (PSTC), in collaboration with its partnering regulatory agencies, has been researching other, potentially more predictive biomarkers of liver injury.3
Analyzing Hepatotoxic Biomarkers With a Multiplex Assay
Multiplex immunoassays enable researchers to measure multiple biomarkers of liver injury at once. The data below describes a Luminex® bead-based multiplex immunoassay (the MILLIPLEX® Rat Liver Injury Panel) that simultaneously measures the following hepatotoxicity biomarkers in the blood:
  • Liver-type arginase 1 (ARG1)
  • α-glutathione-S-transferase (GSTα)
  • 5’nucleotidase (5'NT)
  • Sorbitol dehydrogenase (SDH)
  • Glutamic-oxaloacetic transaminase 1 (GOT1)
These biomarkers are hepatocellular and hepatobiliary enzymes that are released into circulation upon the occurrence of liver injury.4
Performance evaluation of this multiplexed assay panel revealed the robustness of assay sensitivity (MinDC at 12 pg/mL), specificity (no cross-reactivity), accuracy (80 – 100% recovery and linearity), precision (<10% intra-assay CV and <15% inter-assay CV), and wide linear dynamic range (3 logs). The assay panel was biologically qualified using acute acetaminophen- and thioacetamide-induced rat hepatotoxicity models in which centrilobular hepatocyte necrosis was present. Significant changes in blood protein dynamics were observed in both models. Baseline protein levels were barely detectable; hepatocellular enzymes were released into circulation massively upon hepatotoxicity. The peak increase (up to 250-fold) was observed at 24 hours post-dosing. Strongly correlated with protein concentration data, robust increases in enzyme activities were detected. The protein levels gradually declined and returned to normal levels in approximately four days.
The involvement of cytokines in liver injury response was also investigated. Cytokines have been under investigation as potential biomarkers of DILI. A possible mechanism is that molecules released in the early stages of hepatocellular necrosis are recognized by toll-like receptors and thereby lead to the release of pro-inflammatory cytokines.5.6 27 cytokines were measured using the MILLIPLEX® Rat Cytokine/Chemokine Panel, and the dynamic changes found in 8 cytokines paralleled changes in hepatocellular enzyme biomarkers, suggesting that these cytokines were indeed involved in the pathogenesis of hepatotoxicity.
Results of this study demonstrated the robustness and ease of multiplex immunoassays for quantitating multiple biomarkers, particularly for investigators monitoring biomarkers in translational drug-induced liver injury studies.
Materials and Methods
To appraise the performance of the immunoassays, acute acetaminophen (APAP)- and thioacetamide (TAA)-induced rat hepatotoxicity models were established. APAP overdose is the most common cause of acute liver failure in the West. Hepatocellular injury is primarily initiated by APAP metabolite CYP2E1 bioactivation to form reactive intermediates that deplete glutathione and then bind to critical cellular macromolecules. High doses of APAP were shown to induce rat centrilobular hepatocyte necrosis, although less mitochondria dysfunction was found. TAA is a potent hepatotoxicant that undergoes a 2-step bioactivation mediated by microsomal CYP2E1 to thioacetamide sulphoxide (TASO), and further to a reactive metabolite thioacetamide-S, S-dioxide (TASO2). These reactive metabolites may covalently bind to various proteins that also lead to centrilobular hepatocyte necrosis.
Acetaminophen Rat Liver Injury Model
Male Sprague Dawley rats (7 - 8 weeks old) were subjected to fasting conditions overnight and then treated with a single intraperitoneal (IP) injection of acetaminophen in 20% Tween® 80 solution at a dose of 1 g/kg. Before, and approximately 24, 48, and 72 hours after the dose, 250 – 300 μL blood was collected via the sublingual vein under isoflurane anesthetization. 96 hours after the acetaminophen dose, rats were euthanized, and blood samples were collected.
Thioacetamide Rat Liver Injury Model
Male Sprague Dawley rats (7 - 8 weeks old) were subjected to fasting conditions overnight and then treated with a single IP injection of thioacetamide in phosphate-buffered saline (PBS) solution at a dose of 300 mg/kg. Rats were euthanized 24 hours after the dose, and blood samples were collected.
Sample Treatment
Serum or plasma samples were diluted 1:25 in assay buffer and incubated at 37°C overnight (16 - 18 hours).
Multiplex Assays
Assays were conducted using the MILLIPLEX® Rat Liver Injury Panel and the Rat Cytokine/Chemokine Panel following the instructions in the included protocols.
Enzymatic Assay Protocol
The ARG1, GOT1, 5'NT, GSTα, and SDH enzymatic assays were purchased from our company, Crystal Chern, Cayman Chemical, and New York University at Buffalo, respectively, and performed as instructed by the manufacturer’s protocols.
DILI Multiplex Analysis Results
Figure 1 shows the standard curves of each analyte in the panel, on which are plotted the median fluorescence intensities (MFI) corresponding to the biomarker levels in untreated and acetaminophen-treated rat plasma. In all cases, the levels of DILI biomarkers were higher in acetaminophen-treated rats (“TOX”) compared to the untreated rats (“CTRL”).
Graph showing elevated ARG1, GOT1, GSTα, 5’-NT, and SDH levels of blood protein concentration in a rat acetaminophen liver injury model analyzed by the MILLIPLEX® Rat Liver Injury Panel (Cat. No. RLI1MAG-92K).
Figure 1.Blood protein concentrations of ARG1, GOT1, GSTα, 5’-NT, and SDH were elevated in a rat acetaminophen liver injury model. Before (“CTRL”) and 24 hours post-dosing (“TOX”), ~250 μL of blood was collected and plasma samples were prepared from each rat. The MILLIPLEX® Rat Liver Injury Panel was used to quantify blood protein concentrations.
Figure 2 shows the results of enzyme activity assays performed on the plasma samples from untreated and acetaminophen-injected rats. For all five enzyme biomarkers, there was more enzyme activity present per unit sample in the acetaminophen-treated rats.
Graphs showing enzyme activities of ARG1, GOT1, GSTα, 5’-NT, and SDH, which increased correspondingly to acetaminophen dosing.
Figure 2.Enzyme activities of ARG1, GOT1, GSTα, 5’-NT, and SDH increased correspondingly. Before and 24 hours after dosing, ~250 μL of blood was collected and plasma samples were prepared from each rat. ARG1, GOT1, GSTα, 5’-NT, and SDH enzyme activities were measured individually.
Correlated Increases in Hepatotoxicity Biomarkers and Pro-Inflammatory Cytokines
Pro-inflammatory cytokines are released during the early stages of hepatocellular necrosis because toll-like receptors on macrophages recognize certain molecules (DNA fragments, DNA binding proteins, etc.).5 These cytokines cause the release of acute phase proteins, which further affect liver cell injury. To compare the dynamics of cytokine release with increases in hepatotoxicity biomarker levels after acetaminophen treatment, both sets of proteins in blood samples taken at the same time points were measured. The results showed that the 5 hepatotoxicity biomarkers changed with the same dynamics, as did the cytokines MCP-1, GRO/KC/CINC-1, and MIP1α (Figure 3).
Graphs showing the correlation of hepatotoxicity biomarkers and cytokines (MIP1α, EGF, IL-5, MCP-1, GRO/KC/CINC-1, VEGF-A, Fractalkine) in a rat acetaminophen injury model analyzed by the MILLIPLEX® Rat Liver Injury Panel (Cat. No. RLI1MAG-92K) and the MILLIPLEX® Rat Cytokine/Chemokine Panel (Cat. No. RECYTMAG-65K).
Figure 3.Time-course measurements of hepatotoxicity biomarkers and cytokines in a rat acetaminophen injury model. (Left) Hepatotoxicity biomarkers: Before, 24, 48, 72, and 96 hours after dosing, ~250 μL of blood was collected and plasma samples were prepared from each rat. ARG1, GOT1, GSTα, 5’-NT, and SDH blood protein concentrations were measured with MILLIPLEX® Rat Liver Injury Panel. (Right) Cytokines: Before, 24, 48, 72, and 96 hours after dosing, ~250 μL of blood was collected and plasma samples were prepared from each rat. 27 prominent circulating cytokines were measured, of which selected analytes are shown.
Because there are known differences in the tissues affected and molecular mechanisms between different preclinical DILI models, the experiment in Figure 3 was repeated with a thioacetamide-induced rat model of DILI (Figures 4 and 5).7 27 prominent cytokines were measured by the MILLIPLEX® Rat Cytokine/Chemokine Panel and showed distinctive cytokine expression patterns in APAP and TAA models.
Graph showing elevated blood protein concentrations of ARG1, GOT1, GSTα, 5’-NT, and SDH in a rat thioacetamide liver injury model analyzed by the MILLIPLEX® Rat Liver Injury Panel (Cat. No. RLI1MAG-92K).
Figure 4.Blood protein concentrations of ARG1, GOT1, GSTα, 5’-NT, and SDH are elevated in a rat thioacetamide liver injury model. Before and 24 hours after dosing, ~250 μL of blood was collected and plasma samples were prepared from each rat. The MILLIPLEX® Rat Liver Injury Panel was used to quantify blood protein concentration.
Graph showing elevated levels of the cytokines leptin, MIP-1α, IL-10, IL-18, MCP-1, IP-10, GRO, VEGF, Fractalkine, and LIX analyzed by the MILLIPLEX® Rat Cytokine/Chemokine Panel (Cat. No. RECYTMAG-65K).
Figure 5.Leptin, MIP-1α, IL-10, IL-18, MCP-1, IP-10, GRO, VEGF, Fractalkine, and LIX are elevated in a rat thioacetamide liver injury model. Before and 24 hours after dosing, ~250 μL of blood was collected and plasma samples were prepared from each rat. The MILLIPLEX® Rat Cytokine/Chemokine Panel was used to quantify 27 cytokines.
Figures 4 and 5, like Figure 1, show the standard curves for each panel used, on which are plotted the MFI corresponding to biomarker levels in untreated (“CTRL”) and thioacetamide-treated (“TOX”) rat plasma samples. In response to thioacetamide injection, blood protein levels of Leptin, MIP-1α, IL-10, IL-18, MCP-1, IP-10, GRO, VEGF, Fractalkine, and LIX were up-regulated, along with the hepatotoxicity biomarkers.
Conclusion
The MILLIPLEX® Rat Liver Injury and Cytokine/Chemokine panels are quantitative immunoassays that simultaneously and precisely measure circulating hepatotoxicity biomarkers and prominent cytokines. Both assay panels exhibit robust sensitivity, specificity (no cross-activity), accuracy, precision, and wide linear dynamic range.
Acetaminophen and thioacetamide are potent hepatotoxicants that induce centrilobular hepatocyte necrosis in rat. Most liver injury biomarkers are hepatocellular enzymes that have been released into the bloodstream upon injury to the hepatocytes. Acute liver injury is associated with robust and varied immune responses. Changes in levels of circulating cytokines have also been proposed as possible biomarkers. The dynamic changes of hepatotoxicity biomarkers and circulating cytokines were demonstrated in two rat liver injury models. The peak increases were observed 24 hours after drug exposure. Strongly correlated with the protein concentration data, robust increases in enzyme activities were also observed at this time point.
These quantitative immunoassays are a powerful way for investigators to monitor hepatotoxicity in their research.
Materials
Loading
For Research Use Only. Not For Use In Diagnostic Procedures.
References
1.
Chen M, Vijay V, Shi Q, Liu Z, Fang H, Tong W. 2011. FDA-approved drug labeling for the study of drug-induced liver injury. Drug Discovery Today. 16(15-16):697-703. https://doi.org/10.1016/j.drudis.2011.05.007
2.
Njoku D. Drug-Induced Hepatotoxicity: Metabolic, Genetic and Immunological Basis. IJMS. 15(4):6990-7003. https://doi.org/10.3390/ijms15046990
3.
Critical Path Institute. Tucson (AZ): Critical Path Institute, 2008. Predictive Safety Testing Consortium. [Internet]. Available from: http://c-path.org/programs/pstc/pstctools/?anchor=section-578#section-578
4.
Sahini N, Selvaraj S, Borlak J. Whole Genome Transcript Profiling of Drug Induced Steatosis in Rats Reveals a Gene Signature Predictive of Outcome. PLoS ONE. 9(12):e114085. https://doi.org/10.1371/journal.pone.0114085
5.
Jaeschke H, Williams CD, Ramachandran A, Bajt ML. 2012. Acetaminophen hepatotoxicity and repair: the role of sterile inflammation and innate immunity. Liver Int.. 32(1):8-20. https://doi.org/10.1111/j.1478-3231.2011.02501.x
6.
Laverty HG, Antoine DJ, Benson C, Chaponda M, Williams D, Park BK. 2010. The potential of cytokines as safety biomarkers for drug-induced liver injury. Eur J Clin Pharmacol. 66(10):961-976. https://doi.org/10.1007/s00228-010-0862-x
7.
Gad SC. 2008. Preclinical Development Handbook. https://doi.org/10.1002/9780470249055