HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Initial Salt Concentration
![HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Initial Salt Concentration application for HPLC](/deepweb/assets/sigmaaldrich/product/images/207/078/5a100215-3d51-4a23-b923-a4ea2d1093da/800/5a100215-3d51-4a23-b923-a4ea2d1093da.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
(A) 10 mM sodium phosphate buffer, pH 7.5; (B), A + 100 mM NaCl
gradient
15 to 65% B in 60 min
flow rate
0.8 mL/min
column temp.
25 °C
detector
UV, 214 nm
injection
10 μL
sample
MAb-X22, 1.5 mg/mL
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies