Merck
HPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution
HPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-150 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 4.6 mm, 3 μm particle size, stainless steel
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 4.6 mm, 3 μm particle size, stainless steel
CONDITIONS
column
Zenix SEC-150, 30 cm x 4.6 mm I.D., 3 μm, 150 Å particles (Z777016)
column
Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 μm, 300 Å particles (Z777028)
mobile phase
150 mM sodium phosphate, pH 7.0
flow rate
1 mL/min
detector
UV, 214 nm
injection
10 μL
sample
1 mg/mL, IgG
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies