Merck
HPLC Analysis of Ibuprofen Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Ibuprofen Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
Ibuprofen sodium salt
≥98% (GC)
Ibuprofen
meets USP testing specifications
(S)-(+)-Ibuprofen
ReagentPlus®, 99%
Ibuprofen-d3
analytical standard
Ibuprofen
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 20 mM sodium citrate; [B] THF; (90:10, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 225 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC