HPLC Analysis of FMOC-Methionine Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of FMOC-Methionine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/373/852/2ba6f504-d7e6-4a7c-8fdf-0135bce00ad9/800/2ba6f504-d7e6-4a7c-8fdf-0135bce00ad9.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (60:40, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 260 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC