Merck
HPLC Analysis of Fat-Soluble Vitamins on Ascentis® Express C18, Methanol Mobile Phase
HPLC Analysis of Fat-Soluble Vitamins on Ascentis® Express C18, Methanol Mobile Phase application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Retinyl acetate
analytical standard
Retinol palmitate
analytical standard
Cholecalciferol (D3)
analytical standard
Ergocalciferol (D2)
analytical standard
Phylloquinone (K1)
analytical standard
(±)-α-Tocopherol
analytical standard
DL-alpha-Tocopherol acetate
analytical standard
δ-Tocopherol
analytical standard
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
methanol
flow rate
1.5 mL/min
pressure
3249 psi (224 bar)
column temp.
30 °C
detector
UV, 280 nm
injection
5 μL
sample
0.5 mg/mL tocopherol acetate, alpha-tocopherol, delta-tocopherol and K1, 0.05 mg/mL D2, D3 and retinol acetate, 1.5 mg/mL retinol palmitate, all in ethanol.
Description
Analysis Note
Analysis was conducted using standard solution, detection via UV.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany