Merck
HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD, Fast Analysis)
HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD, Fast Analysis) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
standard
Product No.
Description
Pricing
Taurine
≥99%
Caffeine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 3.0 mm I.D., 2.7 μm particles (53967-U)
mobile phase
[A] 100 mM ammoniumformate, pH 3.0 with formic acid; [B] acetonitrile; (15:85, A:B)
flow rate
0.6 mL/min
column temp.
25 °C
detector
ELSD, 55 °C, 3.5 bar nitrogen
injection
2 μL
sample
1000 μg/mL in 10:90, water: acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express HILIC for the efficient separation of caffeine and taurine. Elution is by aqueous normal phase (ANP) as an increase in organic will increase retention.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany