HPLC Analysis of an Energy Drink on Ascentis® Express HILIC, 10 cm (UV Detection)
![HPLC Analysis of an Energy Drink on Ascentis® Express HILIC, 10 cm (UV Detection) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/415/473/98222c61-8c13-45e2-be8c-8884a6e73cb8/800/98222c61-8c13-45e2-be8c-8884a6e73cb8.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 3 mm I.D., 2.7 μm particles (53967-U)
mobile phase
[A] 100 mM ammonium acetate, pH 5 (titrated with acetic acid); [B] water; [C] acetonitrile; (9:1:90, A:B:C:)
flow rate
0.6 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
2 μL
sample
Energy Drink Diluted 1:9 in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany