Merck
GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX B-PM
GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX™ B-PM suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PM Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
Application
Product No.
Description
Pricing
GC Analysis of α-Pinene Enantiomers on Astec® CHIRALDEX B-PM
suitable for GC
standard
Product No.
Description
Pricing
(+)-β-Pinene
analytical standard
(−)-β-Pinene
analytical standard
(−)-β-Pinene
99%
(−)-β-Pinene
≥99%
(−)-β-Pinene
≥97%, FCC, FG
CONDITIONS
column
Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 μm (76023AST)
oven
50 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
0.2 μL, 80:1 split
sample
5 mg/mL in ethanol
peak 1: d-β-pinene
peak 2: l-β-pinene
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC