GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Materials
SPME fiber
SPME fiber assembly Polydimethylsiloxane (PDMS)
df 100 μm(PDMS, needle size 24 ga, for use with manual holderCONDITIONS
sample/matrix
200 ppt each in 2 mL water
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
extraction
15 min immersed in water with rapid stirring
column
SPB-5, 15 m × 0.20 mm I.D., 0.20 μm (24165-U)
oven
120 °C (1 min) to 180 °C at 30 °C/min, then to 290 °C at 10 °C/min
inj. temp.
260 °C (splitless - closed 3 min)
detector
ECD, 300 °C
carrier gas
helium, 37 cm/sec (set at 120 °C)
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany