Merck
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
guard column
Product No.
Description
Pricing
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.10 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
recommended
Product No.
Description
Pricing
Polyimide Sealing Resin
pkg of 5 g
required but not provided
Product No.
Description
Pricing
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 3 ea
standard
Product No.
Description
Pricing
Chlordane (mixture of isomers)
certified reference material, 1000 μg/mL in isooctane
Pesticides Matrix Spike Mix
in methanol (varied), PESTANAL®, analytical standard
Toxaphene solution
certified reference material, 1000 μg/mL in isooctane
Pesticide Surrogate Spike Mix
certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL
EPA CLP Organochlorine Pesticide MIx
certified reference material, 2000 μg/mL each component in hexane: toluene (1:1), ampule of 1 mL
CONDITIONS
column
SPB-608, 30 m x 0.25 mm I.D., 0.25 μm (24103-U)
oven
100 °C (2 min), 15 °C/min to 300 °C (5 min)
inj. temp.
250 °C
detector
ECD, 300 °C
carrier gas
helium, 0.9 ml/min, constant flow
injection
2.0 μL, splitless (0.75 min)
liner
4 mm I.D. single taper
sample
chlorinated pesticide standard (cat. No. 46845-U), 50 ppb in methylene chloride
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany