GC Analysis of Volatiles on SPB®-624, Direct Injection
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SPB-624, 60 m × 0.25 mm I.D., 1.4 μm (24256)
oven
35 °C (4 min), 4 °C/min to 200 °C
inj. temp.
250 °C
scan range
m/z 45-300
carrier gas
helium, 33 cm/sec @ 40 °C
injection
70:1 split
sample
5 ng on column of each component
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany