GC Analysis of Cold Pressed Lemon Essential Oil on Equity-5 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
CONDITIONS
column
Equity-5, 15 m x 0.10 mm I.D., 0.10 μm (28083-U)
oven
75 °C (1 min), 35 °C/min to 200 °C (1 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 45 cm/sec constant
injection
0.10 μL 300:1 split
liner
2 mm I.D., straight
sample
Cold pressed lemon oil, neat
Description
Legal Information
null