Merck
GC Analysis of Cold Pressed Bergamot Essential Oil on SLB®-5ms
GC Analysis of Cold Pressed Bergamot Essential Oil on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C, 3 °C/min to 250 °C (5 min)
inj. temp.
250 °C
detector
FID, 300 °C
carrier gas
helium, 30 cm/sec, constant
injection
Wet needle, 100:1 split
sample
cold pressed bergamot oil
Description
Analysis Note
Bergamot oil is used in cosmetic preparations such as perfumes and lotions, and in flavor formulations included in various candies and beverages. The cold-pressed oil is obtained by the rasping of the peel of the citrus bergamia fruit. The oil is commonly treated prior to use to remove furocoumarins and bergapten, which have the potential to cause advserse skin reactions. In addition, terpenes are often reduced or removed to enhance the desirable top-notes of the oil (1). This application illustrates the analysis of a sample of cold-pressed bergamot oil on the SLB-5ms.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany