Merck
GC Analysis of an Indoor Air Standard on Equity-1
GC Analysis of an Indoor Air Standard on Equity-1 suitable for GC, application for air monitoring
CONDITIONS
column
Equity-1, 30 m × 0.25 mm I.D., 1.0 μm (28049-U)
oven
35 °C (3 min), 8 °C/min to 100 °C, 20 °C/min to 250 °C (10 min)
inj. temp.
250 °C
MSD interface
325 °C
scan range
m/z 33-350
carrier gas
helium, 30 cm/sec @ 35 °C
injection
1 μL, 10:1 split
liner
split, cup design
sample
100 ng on-column of the Japanese Indoor Air Standards Mix (47537-U)
Description
Legal Information
null