Merck
  • [Combination of programmable temperature vaporizer-large volume injection gas chromatography-mass spectrometry and automated mass spectral deconvolution and identification system for the determination of 32 pesticides in fruits and vegetables].
[Combination of programmable temperature vaporizer-large volume injection gas chromatography-mass spectrometry and automated mass spectral deconvolution and identification system for the determination of 32 pesticides in fruits and vegetables].
Se pu = Chinese journal of chromatography (2015-04-24)
Zhaoyun Cao, Renxiang Mou, Li Wu, Lin Xiaoyan, Zhiwei Zhu, Mingxue Chen
ABSTRACT
An analytical method was developed for the simultaneous determination of 32 pesticides including organophosphorus, organochlorine, pyrethroid and carbamate pesticides in fruits and vegetables using gas chromatography-mass spectrometry (GC-MS). The sample was extracted with acetonitrile, and the organic layer was cleaned up with ENVI-Carb and LC-NH2 cartridges. A large volume of 20 µL purified solution was injected into the GC system using programmable temperature vaporizer (PTV). The mass spectrometric detection was operated with full scan mode. The automated mass spectral deconvolution and identification system (AMDIS) and an isotopic internal standard were used for the qualitative and quantitative determination of the 32 pesticides, respectively. The conditions for PTV-large volume injection were studied. Furthermore, the selectivity and durability of the method were also assessed. Under the optimized conditions, the experimental results showed that all the linearities were good within their test ranges, with correlation coefficients more than 0. 995, and the method detection limits of the pesticides were 2.0-5.0 µg/kg. The spiked recoveries (n=6) at three levels in the 0. 010 - 0. 50 mg/kg using spinach, snap bean and cucumber samples as blank matrices were in the range of 65. 2% - 120. 3% with the relative standard deviations (RSDs) varying from 4. 1% to 22. 3%, showing good accuracy of the method. The advantages of the proposed method include high throughput, high sensitivity, reliability, robustness, and the ability to meet the demand of multi-residue pesticide analysis in fruits and vegetables.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS